Learn More
Y15, MedChemExpress
Product Code. 16475168
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475168 10 mg 10mg
16411397 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16475168 Supplier Medchem Express Supplier No. HY12444
MedChemExpress Y15 is a potent and specific inhibitor of focal adhesion kinase (FAK) that inhibits its autophosphorylation activity, decreases the viability of cancer cells, and blocks tumor growth.
Y15 is a potent and specific inhibitor of focal adhesion kinase (FAK) that inhibits its autophosphorylation activity, decreases the viability of cancer cells, and blocks tumor growth.
Specifications
Chemical Name or Material Y15
Physical Form Solid
CAS 4506-66-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C6H14Cl4N4
For Use With (Application) Cancer-Kinase/protease
Synonym FAK Inhibitor 14
Solubility Information H2O : 50 mg/mL (176.05 mM; ultrasonic and warming and heat to 50°C)
SMILES [H]Cl.[H]Cl.[H]Cl.[H]Cl.NC1=CC(N)=C(N)C=C1N
Molecular Weight (g/mol) 284.01
Formula Weight 284.01
Percent Purity 95.0%
Grade Research
Show More Show Less
Research purposes only