Learn More
Tenak™ Yellow Cardboard Cryobox
Product Code. 15260524 Shop All TENAK Products
Change view
Click to view available options
Dimensions (L x W x H):
133 x 133 x 75 mm
136 x 136 x 32 mm
136 x 136 x 50 mm
136 x 136 x 75 mm
133 x 133 x 100 mm
133 x 133 x 130 mm
136 x 136 x 100 mm
136 x 136 x 130 mm
Unit Size:
Pack of 36
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
15260524 133 x 133 x 75 mm Pack of 36
15230614 136 x 136 x 130 mm Pack of 36
15290524 133 x 133 x 130 mm Pack of 36
15240594 136 x 136 x 32 mm Pack of 36
15290594 136 x 136 x 50 mm Pack of 36
15240604 136 x 136 x 75 mm Pack of 36
15200544 133 x 133 x 100 mm Pack of 36
15280604 136 x 136 x 100 mm Pack of 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15260524 Supplier Tenak™ Supplier No. TE22070
Only null left
Add to basket
Edge
Add to basket
Cryobox made of water-repellent cardboard for long-lasting durability.
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Specifications
Color Yellow
Material Cardboard
Shape Square
Type Cryo Box
Dimensions (L x W x H) 133 x 133 x 75 mm