Learn More
Invitrogen™ YES1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580243
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human U20S whole cell, human CACO-2 whole cell, human A431 whole cell, human HepG2 whole cell, human MCF-7 whole cell, human PC-3 whole cell. IHC: human mammary cancer tissue.
YES1 is a recombinant human protein kinase that is a member of the SRC family of non-receptor tyrosine kinases and is the cellular homolog of the Yamaguchi sarcoma virus oncogene. It is involved in the regulation of cell growth and survival, including roles in apoptosis, cell-cell adhesion, cytoskeleton remodeling, and differentiation.
Specifications
| YES1 | |
| Polyclonal | |
| Unconjugated | |
| Yes1 | |
| cellular yes-1 protein; c-Src; c-Yes; EC 2.7.10.2; HsT441; p60c-yes; p60-Src; P61-YES; pp60c-src; Proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; tyrosine-protein kinase Yes; viral oncogene yes homolog; v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1; Yamagichi sarcoma viral (v-yes-1) oncogene homolog 1; Yamaguchi sarcoma oncogene; Yamaguchi sarcoma viral (v-yes) oncogene homolog 1; Yamaguchi sarcoma viral oncogene homolog 1; Yes; YES proto-oncogene 1, Src family tyrosine kinase; Yes1; YES1 proto-oncogene, Src family tyrosine kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7525 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P07947 | |
| Yes1 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human Yes1 (7-35aa KENKSPAIKYRPENTPEPVSTSVSHYGAE). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.