missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Yhhu 3792

Description
Yhhu 3792 is a notch signaling pathway activator; enhances the self-renewal capability of neural stem cells (NSCs) via Notch signaling pathway activation in vitro and in vivo. Promotes the expression of Notch target genes, Hes3 and Hes5. Expands the NSCs pool and promotes endogenous neurogenesis in the hippocampal dentate gyrus (DG) after chronic administration in mice. Increases the spatial and episodic memory in mice.
Specifications
Specifications
| Chemical Name or Material | 5-(3-Methoxyphenoxy)-N2-[4-(1-methylethyl)phenyl]-2,4-quinazolinediamine hydrochloride |
| CAS | 2624336-93-0 |
| Quantity | 10 mg |
| Target | Compounds for NSC Proliferation and Renewal |
| Molecular Formula | C24H24N4O2.HCl |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?