missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YIF1B Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18478AF488
This item is not returnable.
View return policy
Description
YIF1B Polyclonal antibody specifically detects YIF1B in Human samples. It is validated for Western Blot
Specifications
| YIF1B | |
| Polyclonal | |
| Western Blot | |
| FinGER8, FLJ23477, FLJ50074, MGC39135, protein YIF1B, Yip1 interacting factor homolog B (S. cerevisiae), YIP1-interacting factor homolog B | |
| Recognizes region between residues Between between 1 and 50.. | |
| 0.1 mL | |
| Neuroscience | |
| 90522 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction