missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ YIL 781 hydrochloride

Description
YIL 781 hydrochloride is a ghrelin receptor antagonist (GHS-R1a) (Ki = 17 nM). Displays no significant affinity for the motilin receptor (Ki = 6 μM). Blocks the effects of ghrelin on insulin secretion both in vivo and in vitro. Improves glucose homeostasis in vivo.
Specifications
Specifications
| Chemical Name or Material | 6-(4-Fluorophenoxy)-2-methyl-3-[[(3S)-1-(1-methylethyl)-3-piperidinyl]methyl]-4(3H)-quinazolinone hydrochloride |
| CAS | 875258-85-8 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C24H28FN3O2.HCl |
| Formula Weight | Observed MW: 445.96 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?