Learn More
Invitrogen™ YIPF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113798
Description
Immunogen sequence: DMMQPQQTYT GQIYQPTQAY PPTTPQPFYG DSFEEEPPLL EELGINFDHI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 100%.
Plays a role in transport between endoplasmic reticulum and Golgi.
Specifications
| YIPF5 | |
| Polyclonal | |
| Unconjugated | |
| Yipf5 | |
| 2610311I19Rik; AA408236; Ac2-256; FinGER5; Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5; golgi membrane protein SB140; PP12723; Protein YIPF5; protein YIPF5-like protein; SB140; SMAP5; SMAP-5; smooth muscle cell associated protein 5; smooth muscle cell-associated protein 5; UNQ3123/PRO10275; Yip1 domain family member 5; Yip1 domain family, member 5; YIP1 family member 5; Yip1a; Yipf5; YPT-interacting protein 1 A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 67180 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9EQQ2 | |
| Yipf5 | |
| Immunogen sequence: DMMQPQQTYT GQIYQPTQAY PPTTPQPFYG DSFEEEPPLL EELGINFDHI (aa 54-103). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.