missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ YIPF5 Recombinant Protein Antigen
Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Beschreibung
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human YIPF5. Source: E.coli Amino Acid Sequence: NLNTDFYQTSYSIDDQSQQSYDYGGSGGPYSKQYAGYDYSQQGRFVPPDMMQPQQPYTGQIYQPTQAYTPASPQPFYGNNFEDEPPLLE The YIPF5 Recombinant Protein Antigen is derived from E. coli. The YIPF5 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Spezifikation
Spezifikation
| Gene ID (Entrez) | 81555 |
| Purification Method | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | YIPF5 Recombinant Protein Antigen |
| Content And Storage | Store at −20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Alias | FinGER5, Golgi Membrane Protein SB140, Protein YIPF5, SB140, SMAP5, SMAP-5, Smooth Muscle Cell Associated Protein 5, Smooth Muscle Cell-Associated Protein 5, Yip1 Domain Family Member 5, Yip1 Domain Family, Member 5, YIP1 Family Member 5, YIP1A, YPT-Inter |
| Gene Symbol | YIPF5 |
| Label Type | Unlabeled |
| Product Type | Recombinant Protein Antigen |
| Mehr anzeigen |
For Research Use Only.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur