missing translation for 'onlineSavingsMsg'
Learn More

YKL-05-099, MedChemExpress

Product Code. 16400739
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400739

Brand: Medchem Express HY101147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress YKL-05-099 is a salt-inducible kinase (SIK) inhibitor. YKL-05-099 binds to SIK1 and SIK3 with IC50s of ∼10 and ∼30 nM, respectively. YKL-05-099 has slightly less potent SIK2-inhibitory (IC50=40 nM).

YKL-05-099 is a salt-inducible kinase (SIK) inhibitor. YKL-05-099 binds to SIK1 and SIK3 with IC50s of ∼10 and ∼30 nM, respectively. YKL-05-099 has slightly less potent SIK2-inhibitory (IC50=40 nM).

Specifications

Chemical Name or Material YKL-05-099
CAS 1936529-65-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C32H34ClN7O3
Quantity 1 mg
Solubility Information DMSO : ≥ 75 mg/mL (124.98 mM)
SMILES O=C1N(C2=NC=C(OC)C=C2)C3=NC(NC4=CC=C(C5CCN(C)CC5)C=C4OC)=NC=C3CN1C6=C(C)C=CC=C6Cl
Molecular Weight (g/mol) 600.11
Formula Weight 600.11
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.