missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ YKT6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16538721
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16538721 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16538721 Supplier Invitrogen™ Supplier No. PA595879

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EFSKQVDRID WPVGSPATIH YPALDGHLSR YQNPREADPM TKVQAELDET KIILHNTMES LLERGEKLDD LVSKSEVLGT QSKAFYKTAR KQNSCCAIM.

This gene product is one of the SNARE recognition molecules implicated in vesicular transport between secretory compartments. It is a membrane associated, isoprenylated protein that functions at the endoplasmic reticulum-Golgi transport step. This protein is highly conserved from yeast to human and can functionally complement the loss of the yeast homolog in the yeast secretory pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YKT6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene YKT6
Gene Accession No. Q5EGY4
Gene Alias 0610042I15Rik; 1810013M05Rik; AW105923; prenylated SNARE protein; prenylated SNARE protein (Ykt6); R-SNARE; SNARE protein Ykt6; synaptobrevin homolog YKT6; Ykt6; YKT6 homolog; YKT6 homolog (S. Cerevisiae); YKT6 v-SNARE homolog (S. cerevisiae); YKT6 v-SNARE protein; YKT6, S. cerevisiae, homolog of
Gene Symbols YKT6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-198 of human YKT6 (NP_0065461).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64351
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.