missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YLAT2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143051
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
Involved in the sodium-independent uptake of dibasic amino acids and sodium-dependent uptake of some neutral amino acids. Requires coexpression with SLC3A2/4F2hc to mediate the uptake of arginine, leucine and glutamine. Also acts as an arginine/glutamine exchanger, following an antiport mechanism for amino acid transport, influencing arginine release in exchange for extracellular amino acids. Plays a role in nitric oxide synthesis in human umbilical vein endothelial cells (HUVECs) via transport of L-arginine. Involved in the transport of L-arginine in monocytes. Reduces uptake of ornithine in retinal pigment epithelial (RPE) cells. [UniProt].
Specifications
| YLAT2 | |
| Polyclonal | |
| Unconjugated | |
| SLC7A6 | |
| AI643885; amino acid permease; cationic amino acid transporter, y+ system; KIAA0245; LAT-2; LAT3; Slc7a6; solute carrier family 7 (amino acid transporter light chain, y+L system), member 6; solute carrier family 7 (cationic amino acid transporter, y+ system), member 6; solute carrier family 7 member 6; solute carriersolute carrier family 7 (cationic amino acid transporter, y+ system), member 6; y(+)L-type amino acid transporter 2; Y+L amino acid transporter 2; Y+LAT2; y+LAT-2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9057 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q92536 | |
| SLC7A6 | |
| Peptide with sequence C-ELDVAEEKKDERKTD. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction