Learn More
YLF-466D, MedChemExpress
Product Code. 16492927
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16492927
Brand: Medchem Express HY15840
MedChemExpress YLF-466D is a newly developed AMPK activator, which inhibits platelet aggregation.
YLF-466D is a newly developed AMPK activator, which inhibits platelet aggregation.
Specifications
Chemical Name or Material YLF-466D
Physical Form Solid
CAS 1273323-67-3
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C29H20ClNO3
For Use With (Application) Cancer-Kinase/protease
Synonym C24
Solubility Information DMSO : ≥ 100 mg/mL (214.62 mM)
SMILES O=C(O)C1=CC=CC(CN2C(/C(C3=C2C=CC=C3)=C(C4=CC=C(Cl)C=C4)\C5=CC=CC=C5)=O)=C1
Molecular Weight (g/mol) 465.93
Formula Weight 465.93
Percent Purity 98.41%
Grade Research
Show More Show Less
Research purposes only