missing translation for 'onlineSavingsMsg'
Learn More

YM-58483, MedChemExpress

Product Code. 16439736
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439736

Brand: Medchem Express HY100831

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress YM-58483 (BTP2) is the first selective and potent inhibitor of CRAC channels and subsequent Ca2+ signals. YM-584832 is a blocker of store-operated Ca2+ entry (SOCE).

YM-58483 (BTP2) is the first selective and potent inhibitor of CRAC channels and subsequent Ca2+ signals. YM-584832 is a blocker of store-operated Ca2+ entry (SOCE).

Specifications

Chemical Name or Material YM-58483
Physical Form Solid
CAS 223499-30-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H9F6N5OS
For Use With (Application) COVID-19-immunoregulation
Synonym BTP2
Solubility Information DMSO : 125 mg/mL (296.69 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N=NS1)NC2=CC=C(N3N=C(C(F)(F)F)C=C3C(F)(F)F)C=C2
Molecular Weight (g/mol) 421.32
Formula Weight 421.32
Percent Purity 98.75%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.