missing translation for 'onlineSavingsMsg'
Learn More

YO-01027, MedChemExpress

Product Code. 16480196
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16480196

Brand: Medchem Express HY13526

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress YO-01027 (Dibenzazepine;DBZ) is a potent γ-secretase inhibitor with IC50 values of 2.92 and 2.64 nM for Notch and APPL cleavage, respectively.

YO-01027 (Dibenzazepine

Specifications

Chemical Name or Material YO-01027
CAS 209984-56-5
Color White
Purity Grade Notes Research
Molecular Formula C26H23F2N3O3
Quantity 2 mg
Synonym Dibenzazepine DBZ
Solubility Information DMSO : 50 mg/mL (107.88 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(N[C@@H](C)C(N[C@H]1C2=CC=CC=C2C3=CC=CC=C3N(C)C1=O)=O)CC4=CC(F)=CC(F)=C4
Molecular Weight (g/mol) 463.48
Formula Weight 463.48
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.