Learn More
MP Biomedicals™ YPD (YEPD) Broth ( Powder)
Product Code. p-4474919 Shop All MP Biomedicals Products
Click to view available options
Quantity:
454 g
1000 g
Unit Size:
1kg
454g
Product Code. 11347749
Brand: MP Biomedicals™ 114001032
Rich medium for the growth of S. cerevisiae in liquid medium
  • Contains, dextrose, peptone, and yeast extract
  • pH 6.5 at 300°C
  • Bulk powder packaging
Formulation
  • 50g/L
  • Contents/liter: 20g peptone, 10g yeast extract, 20g dextrose
Yeast Cell Culture
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type Microbiology Media
Formulation Casein Peptone type-M 20g/L, Yeast extract 10g/L, Dextrose 20g/L
For Use With (Application) For growth of Saccharomyces cerevisiae in liquid medium
Packaging Type Bulk Powder
Quantity 1000 g