missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YPEL1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-41163CL1
This item is not returnable.
View return policy
Description
YPEL1 Polyclonal antibody specifically detects YPEL1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| YPEL1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| DiGeorge syndrome-related protein, MGC64992, protein yippee-like 1, yippee (Drosophila) homolog-like 1, yippee-like 1 (Drosophila) | |
| Antibody was raised against a 11 amino acid synthetic peptide near the amino terminus of human YPEL1. The immunogen is located within the first 50 amino acids of YPEL1. Amino Acid Squence: AAVKMTKSKTFQA | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 29799 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction