missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YPEL2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41164B
This item is not returnable.
View return policy
Description
YPEL2 Polyclonal antibody specifically detects YPEL2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| YPEL2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Biotin | |
| DiGeorge syndrome-related protein, Protein Yippee-Like 2, Yippee-Like 2 (Drosophila) | |
| Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human YPEL2. The immunogen is located within amino acids 30 - 80 of YPEL2. Amino Acid Squence: SNPMPQTGEVYPAPGS | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 388403 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction