missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ YTHDC2 Recombinant Rabbit Monoclonal Antibody (HL1862)

Product Code. 17809665
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17809665 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17809665 Supplier Invitrogen™ Supplier No. MA547205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YTHDC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1862
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene YTHDC2
Gene Accession No. B2RR83, Q9H6S0
Gene Alias 3010002F02Rik; 3'-5' RNA helicase YTHDC2; BC037178; CAHL; CsA-associated helicase-like protein; DKFZp564A186; FLJ10053; FLJ2194; hYTHDC2; Keen to exit meiosis leaving testes under-populated protein; Ketu; mYTHDC2; probable ATP-dependent RNA helicase YTHDC2; YTH domain containing 2; YTH domain-containing protein 2; YTH domain-containing protein C2; Ythdc2
Gene Symbols YTHDC2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human YTHDC2. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240255, 64848
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.