missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YTHDC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557920
This item is not returnable.
View return policy
Description
Immunogen sequence: APSKPWSQVD EATIRAIIAV LSTEEQSAGL QQPSGIGQRP RPMSSEELPL ASSWRSNNSR KSSADTEFSD ECTTAERVLM KSPSPALHP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 97%.
Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability.
Specifications
| YTHDC2 | |
| Polyclonal | |
| Unconjugated | |
| YTHDC2 | |
| 3010002F02Rik; 3'-5' RNA helicase YTHDC2; BC037178; CAHL; CsA-associated helicase-like protein; DKFZp564A186; FLJ10053; FLJ2194; hYTHDC2; Keen to exit meiosis leaving testes under-populated protein; Ketu; mYTHDC2; probable ATP-dependent RNA helicase YTHDC2; YTH domain containing 2; YTH domain-containing protein 2; YTH domain-containing protein C2; Ythdc2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64848 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H6S0 | |
| YTHDC2 | |
| Recombinant protein corresponding to Human YTHDC2. Recombinant protein control fragment (Product #RP-96802). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction