missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YTHDF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570854
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs, and regulates mRNA stability. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability. Acts as a regulator of mRNA stability: binding to m6A-containing mRNAs results in the localization to mRNA decay sites, such as processing bodies (P-bodies), leading to mRNA degradation. Also acts as a promoter of cap-independent mRNA translation following heat shock stress: upon stress, relocalizes to the nucleus and specifically binds mRNAs with some m6A methylation mark at their 5'-UTR, protecting demethylation of mRNAs by FTO, thereby promoting cap-independent mRNA translation.
Specifications
| YTHDF2 | |
| Polyclonal | |
| Unconjugated | |
| YTHDF2 | |
| 9430020E02Rik; CAHL; CLL-associated antigen KW-14; HGRG8; high glucose-regulated protein 8; high-glucose-regulated protein 8; LOW QUALITY PROTEIN: YTH domain-containing family protein 2; NY-REN-2; renal carcinoma antigen NY-REN-2; YTH domain family 2; YTH domain family protein 2; YTH domain family, member 2; YTH domain-containing family protein 2; YTH N(6)-methyladenosine RNA binding protein 2; YTH N6-methyladenosine RNA binding protein 2; yth2; YTHDF2; zgc:56224 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51441 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y5A9 | |
| YTHDF2 | |
| synthetic peptide directed towards the C-terminal region of human YTHDF2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction