Learn More
Thermo Scientific™ Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form
Product Code. 15463835
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
Diameter (Metric) Outer:
51mm
62mm
77mm
Height (Metric):
58mm
72mm
90mm
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Outer Height (Metric) unitSize
15463835 100 mL 62mm 72mm Each
15453835 50 mL 51mm 58mm Each
15473835 200 mL 77mm 90mm Each
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15463835
Brand: Thermo Scientific™ 041290.KF
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Material Zirconium Oxide
Quantity 1 Piece
Capacity (Metric) 100 mL
Diameter (Metric) Outer 62mm
Height (Metric) 72mm
Product Type Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form
Diameter (Metric) Base 41mm