Learn More
(Z)-rac-Luliconazole, TRC
Specifications
Specifications
| Chemical Name or Material | (Z)-rac-Luliconazole |
| CAS | 101529-76-4 |
| Molecular Formula | C14H9Cl2N3S2 |
| Quantity | 10 mg |
| Synonym | (Z)-alpha-[4-(2,4-Dichlorophenyl)-1,3-dithiolan-2-ylidene]-1H-imidazole-1-acetonitrile |
| InChI Formula | InChI=1S/C14H9Cl2N3S2/c15-9-1-2-10(11(16)5-9)13-7-20-14(21-13)12(6-17)19-4-3-18-8-19/h1-5,8,13H,7H2/b14-12- |
| SMILES | Clc1ccc(C2CS\C(=C(/C#N)\n3ccnc3)\S2)c(Cl)c1 |
| IUPAC Name | (2Z)-2-[4-(2,4-dichlorophenyl)-1,3-dithiolan-2-ylidene]-2-imidazol-1-ylacetonitrile |
| Molecular Weight (g/mol) | 354.28 |
| Formula Weight | 352.9615 |