Learn More
LNI Swissgas ZA RACK FID 4U 15
Shop All LNI Swissgas Products
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Description
- Zero air generator RACK 2U for FID 15 L/min.
Specifications
Specifications
| Voltage | 100/115/220/230 V |
| Type | Zero Air Gas Generator |