Learn More
LNI Swissgas ZA TOTAL C 3.0
Shop All LNI Swissgas Products
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Description
- Zero air generator 3.0 L/min HC/carbon dioxide/moisture removal with integrated compressor.
Specifications
Specifications
| Voltage | 100/115/220/230 V |
| Type | Zero Air Gas Generator |