missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Zacopride hydrochloride
GREENER_CHOICE

Product Code. p-4784842
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18095452

Brand: Tocris Bioscience™ 1795/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent 5-HT3 antagonist; also 5-HT4 agonist

Highly potent 5-HT3 receptor antagonist (Ki = 0.38 nM) and 5-HT4 receptor agonist (Ki = 373 nM). Antiemetic and anxiolytic following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101303-98-4
Molecular Formula C15H21Cl2N3O2

Specifications

Quantity 10 mg
Formula Weight 346.26
Purity >99%
Molecular Weight (g/mol) 346.252
Chemical Name or Material Zacopride hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.