missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CtBP1 Monoclonal specifically detects CtBP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | CtBP1 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 4D6 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | BARS, brefeldin A-ribosylated substrate, CTBP, CtBP1, C-terminal binding protein 1, C-terminal-binding protein 1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.158, MGC104684 |
| Gene Symbols | CTBP1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?