missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAG Antibody (Zagy-1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80083AF350
This item is not returnable.
View return policy
Description
ZAG Monoclonal antibody specifically detects ZAG in Human samples. It is validated for Western Blot, ELISA
Specifications
| ZAG | |
| Monoclonal | |
| Alexa Fluor 350 | |
| alpha-2-glycoprotein 1, zinc, alpha-2-glycoprotein 1, zinc-binding, ZA2G, ZAGzn-alpha-2-GP, zinc, zinc-alpha-2-glycoprotein, zn-alpha-2-glycoprotein, ZNGP1 | |
| Recombinant human ZAG. | |
| 0.1 mL | |
| Endocrinology, Immunology, Innate Immunity, Signal Transduction | |
| 563 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| Zagy-1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction