missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 214.00 - € 584.00
Specifications
| Antigen | ZAK |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227332
|
Novus Biologicals
NBP3-35439-20ul |
20 μL |
€ 214.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232331
|
Novus Biologicals
NBP3-35439-100ul |
100 μL |
€ 584.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZAK Polyclonal antibody specifically detects ZAK in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| ZAK | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 51776 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| AZK, EC 2.7.11, EC 2.7.11.25, HCCS-4, Human cervical cancer suppressor gene 4 protein, Leucine zipper- and sterile alpha motif-containing kinase, mitogen-activated protein kinase kinase kinase MLT, mixed lineage kinase 7, mixed lineage kinase with a leucine zipper and a sterile alpha motif, Mixed lineage kinase-related kinase, mixed lineage kinase-related kinase MRK-beta, MLK7, mLklak, MLK-like mitogen-activated protein triple kinase, MLK-related kinase, MLT, MLTK, MRKcervical cancer suppressor gene 4 protein, pk, protein kinase, sterile alpha motif and leucine zipper containing kinase AZK, Sterile alpha motif- and leucine zipper-containing kinase AZK | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-312 of human ZAK (NP_598407.1).,, Sequence:, EEMDMDHIMTWATDVAKGMHYLHMEAPVKVIHRDLKSRNVVIAADGVLKICDFGASRFHNHTTHMSLVGTFPWMAPEVIQSLPVSETCDTYSYGVVLWEMLTREVPFKGLEGLQVAWLVVEKNERLTIPSSCPRSFAELLHQCWEADAKKRPSFKQIISILESMSNDTSLPDKCNSFLHNKAEWRCEIEATLERLKKLERDLSFKEQELKERE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title