missing translation for 'onlineSavingsMsg'
Learn More

Zaltoprofen, MedChemExpress

Product Code. 16438396
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16438396

Brand: Medchem Express HYB0619

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zaltoprofen (CN100), a non-steroidal anti-inflammatory drug (NSAID), is a preferential and orally active COX-2 inhibitor, with IC50s of 1.3 and 0.34 μM for COX-1 and COX-2, respectively. Zaltoprofen exhibits powerful anti-inflammatory effects as well as an analgesic action on inflammatory pain.

Zaltoprofen (CN100), a non-steroidal anti-inflammatory drug (NSAID), is a preferential and orally active COX-2 inhibitor, with IC50s of 1.3 and 0.34 μM for COX-1 and COX-2, respectively. Zaltoprofen exhibits powerful anti-inflammatory effects as well as an analgesic action on inflammatory pain.

Specifications

Chemical Name or Material Zaltoprofen
Physical Form Solid
CAS 74711-43-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H14O3S
For Use With (Application) Neuroscience-Neuromodulation
Synonym CN100
Solubility Information DMSO : ≥ 100 mg/mL (335.17 mM)
SMILES CC(C1=CC=C(SC2=C3C=CC=C2)C(CC3=O)=C1)C(O)=O
Molecular Weight (g/mol) 298.36
Formula Weight 298.36
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.