Learn More
Zaltoprofen, TRC
Specifications
Specifications
| Chemical Name or Material | Zaltoprofen |
| CAS | 74711-43-6 |
| Molecular Formula | C17 H14 O3 S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C17H14O3S/c1-10(17(19)20)11-6-7-15-12(8-11)9-14(18)13-4-2-3-5-16(13)21-15/h2-8,10H,9H2,1H3,(H,19,20) |
| SMILES | CC(C(=O)O)c1ccc2Sc3ccccc3C(=O)Cc2c1 |
| IUPAC Name | 2-(6-oxo-5H-benzo[b][1]benzothiepin-3-yl)propanoic acid |
| Molecular Weight (g/mol) | 298.36 |
| Formula Weight | 298.0664 |