missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zalutumumab Recombinant Human Monoclonal Antibody (2F8 (Zalutumumab; HuMax-EGFR))
GREENER_CHOICE

Product Code. 30087868
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30087868 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30087868 Supplier Invitrogen™ Supplier No. MA551874

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

This antibody binds domain III of the human epidermal growth factor receptor (EGFR).

Zalutumumab is a fully human IgG1 monoclonal antibody designed to bind with selectivity to the epidermal growth factor receptor (EGFR). Zalutumumab has been investigated for the treatment of Squamous Cell Cancer and Head and Neck Cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zalutumumab
Applications ELISA, Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone 2F8 (Zalutumumab; HuMax-EGFR)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias HuMax-EGFr
Host Species Human
Immunogen The original monoclonal antibody was generated by immunizing HuMAb mice with alternating A431 cells and purified EGFR administration.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.