missing translation for 'onlineSavingsMsg'
Learn More

Zanamivir Azide Triacetate Methyl Ester, TRC

Product Code. 30456462
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456462 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456462 Supplier Toronto Research Chemicals Supplier No. TRCZ1480402MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zanamivir Azide Triacetate Methyl Ester
CAS 130525-58-5
Molecular Formula C18H24N4O10
Quantity 2 mg
Synonym 5-(Acetylamino)-2,6-anhydro-4-azido-3,4,5-trideoxy-D-glycero-D-galacto-non-2-enonic Acid Methyl Ester 7,8,9-Triacetate
InChI Formula InChI=1S/C18H24N4O10/c1-8(23)20-15-12(21-22-19)6-13(18(27)28-5)32-17(15)16(31-11(4)26)14(30-10(3)25)7-29-9(2)24/h6,12,14-17H,7H2,1-5H3,(H,20,23)/t12-,14+,15+,16+,17+/m0/s1
SMILES COC(=O)C1=C[C@H](N=[N+]=[N-])[C@@H](NC(=O)C)[C@@H](O1)[C@H](OC(=O)C)[C@@H](COC(=O)C)OC(=O)C
IUPAC Name methyl (2R,3R,4S)-3-acetamido-4-azido-2-[(1S,2R)-1,2,3-triacetyloxypropyl]-3,4-dihydro-2H-pyran-6-carboxylate
Molecular Weight (g/mol) 456.4
Formula Weight 456.1492
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.