missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAP70 Antibody (2F3.2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47774F
This item is not returnable.
View return policy
Description
ZAP70 Monoclonal antibody specifically detects ZAP70 in Human samples. It is validated for Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| ZAP70 | |
| Monoclonal | |
| FITC | |
| 70 kDa zeta-associated protein, EC 2.7.10, EC 2.7.10.2, SRKFLJ17679, STDFLJ17670, Syk-related tyrosine kinase, tyrosine-protein kinase ZAP-70, TZK, ZAP-70, zeta-chain (TCR) associated protein kinase (70 kD), zeta-chain (TCR) associated protein kinase 70kDa, zeta-chain associated protein kinase, 70kD | |
| Recombinant ZAP-70 protein including residues 1-254 and encompassing SH2 domains of human ZAP70 (Uniprot: P43403) | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 7535 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Simple Western, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2F3.2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction