Learn More
Invitrogen™ ZAR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106806
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZAR1.
This maternal effect gene is oocyte-specific and encodes a protein that is thought to function in the initiation of embryogenesis. A similar protein in mouse is required for female fertility. [provided by RefSeq, Jul 2013].
Specifications
| ZAR1 | |
| Polyclonal | |
| Unconjugated | |
| ZAR1 | |
| oocyte-specific maternal effect factor; Z3CXXC6; ZAR1; zinc finger, 3CxxC-type 6; zygote arrest 1; zygote arrest protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 326340 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86SH2 | |
| ZAR1 | |
| A synthesized peptide derived from human ZAR1(Accession Q86SH2), corresponding to amino acid residues Q238-G288. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |