Learn More
IGSF11 Monoclonal Antibody (OTI3A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803775S
Encompass_Preferred
 View more versions of this product
Product Code. 17573003
  • € 242.00 / 30µL
Description
IGSF11 Monoclonal antibody specifically detects IGSF11 in Human samples. It is validated for Western Blot
Specifications
Specifications
IGSF11
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O14713
Mouse
Affinity chromatography
RUO
9270
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI3A4
Unconjugated
ITGB1BP1
1700025L02Rik; brain and testis-specific immunoglobin superfamily protein; brain and testis-specific immunoglobulin superfamily protein; BTIGSF; Bt-IGSF; cancer/testis antigen 119; CT119; CXADR like 1; CXADRL1; Igsf11; Igsf13; Immunoglobulin superfamily member 11; immunoglobulin superfamily, member 11; immunoglobulin superfamily, member 13; V-set and immunoglobulin domain containing 3; V-set and immunoglobulin domain-containing protein 3; VSIG3
Full length human recombinant protein of human ITGB1BP1 produced in E.coli.
30 μL