missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otubain-2 Antibody (OTI11B3), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73172B
This item is not returnable.
View return policy
Description
Otubain-2 Monoclonal specifically detects Otubain-2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Otubain-2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| OTUB2 | |
| Full length recombinant protein of human Otubain-2 (NP_075601) produced in HEK293T cells. | |
| 0.1 mL | |
| 78990.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI11B3 | |
| Western Blot, Flow Cytometry | |
| C14orf137chromosome 14 open reading frame 137, Deubiquitinating enzyme OTUB2, EC 3.4.19.12, FLJ21916, MGC3102, OTB2otubain-2, OTU domain, ubiquitin aldehyde binding 2, OTU domain-containing ubiquitin aldehyde-binding protein 2, OTU2ubiquitin thioesterase OTUB2, Otubain-2, ubiquitin-specific protease otubain 2, Ubiquitin-specific-processing protease OTUB2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction