missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBED4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560980
This item is not returnable.
View return policy
Description
Immunogen sequence: PPDSLERMDF KSEQEDMKQT DSGGERAGLG GTGCSCKPPG KYLSAESEDD YGALFSQYSS TLYDVAMEAV TQSLLSSRNM SSRKKSPAWK HFFISPRDST KAICMYCVKE FSRGKNEKDL STSCLMRHVR RAHPTVLIQ Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
Specifications
| ZBED4 | |
| Polyclonal | |
| Unconjugated | |
| ZBED4 | |
| 1700009F06Rik; KIAA0637; Zbed4; Zinc finger BED domain-containing protein 4; zinc finger BED-type containing 4; zinc finger, BED domain containing 4; zinc finger, BED type containing 4; zinc finger, BED-type containing 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 223773, 315211, 9889 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75132, Q80WQ9 | |
| ZBED4 | |
| Recombinant protein corresponding to Human ZBED4. Recombinant protein control fragment (Product #RP-91362). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction