missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76970-0.025mg
This item is not returnable.
View return policy
Description
ZBTB1 Polyclonal specifically detects ZBTB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ZBTB1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| zinc finger and BTB domain containing 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 22890 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| AAH50719 | |
| ZBTB1 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the center of human ZBTB1. The immunogen is located within amino acids 260 - 310 of ZBTB1. Amino Acid Squence: SAQTDKYRGDTSQA | |
| 0.025 mg | |
| Primary | |
| At least two isoforms of ZBTB1 are known to exist. This ZBTB1 antibody will not cross-react with ZBTB2. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction