missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB17 Monoclonal Antibody (OTI7D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811343
This item is not returnable.
View return policy
Description
Transcription factor that can function as an activator or repressor depending on its binding partners, and by targeting negative regulators of cell cycle progression. Plays a critical role in early lymphocyte development, where it is essential to prevent apoptosis in lymphoid precursors, allowing them to survive in response to IL7 and undergo proper lineage commitment. Has been shown to bind to the promoters of adenovirus major late protein and cyclin D1 and activate transcription. Required for early embryonic development during gastrulation.Specifications
| ZBTB17 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13105 | |
| ZBTB17 | |
| Human recombit protein fragment corresponding to amino acids 112-457 of human ZBTB17 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7D8 | |
| Unconjugated | |
| ZBTB17 | |
| MIZ-1, ZNF151, ZNF60, pHZ-67 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7709 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction