missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB17 Monoclonal Antibody (OTI7D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811343S
This item is not returnable.
View return policy
Description
ZBTB17 Monoclonal antibody specifically detects ZBTB17 in Human samples. It is validated for Western BlotSpecifications
| ZBTB17 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P67812 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23478 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7D8 | |
| Unconjugated | |
| SEC11A | |
| AA589413; LP-1; Miz1; Miz-1; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; mZ13; pHZ-67; Polyomavirus late initiator promoter-binding protein; ZBTB17; Zfp100; Zfp-100; zinc finger and BTB domain containing 17; zinc finger and BTB domain-containing protein 17; Zinc finger protein 100; zinc finger protein 151; zinc finger protein 151 (pHZ-67); zinc finger protein 60; Zinc finger protein Z13; ZNF151; ZNF60 | |
| Full length human recombinant protein of human SEC11A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction