Learn More
ZBTB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP1-88787
 View more versions of this product
Product Code. 18739123
  • ONLINE EXCLUSIVE PRICE
    € 539.00
    € 509.25 / 0.10mL
    Save € 29.75
    5% Off
Description
ZBTB2 Polyclonal specifically detects ZBTB2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZBTB2
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Polyclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.