Learn More
ZBTB2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | ZBTB2 |
|---|---|
| Concentration | 350 μg/mL |
| Applications | Immunoprecipitation, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18019675
|
Proteintech
26436-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18064505
|
Proteintech
26436-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ZBTB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZBTB2. This antibody recognizes human antigen. The ZBTB2 antibody has been validated for the following applications: IP, ELISA analysis.
Specifications
| ZBTB2 | |
| Immunoprecipitation, ELISA | |
| Rabbit | |
| 57621 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 350 μg/mL | |
| Liquid | |
| Human | |
| KIAA1483, ZBTB2, ZNF437 | |
| Primary |