missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB20 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23987-1-AP-150UL
This item is not returnable.
View return policy
Description
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZBTB20 (zinc finger and BTB domain containing 20), also known as HOF, DPZF, ODA-8S or ZNF288, is a 741 amino acid protein that localizes to the nucleus and contains one BTB (POZ) domain and five C2H2-type zinc fingers. Expressed in thymus, spleen, lymph node and fetal liver, ZBTB20 exists as either a monomer or a homodimer that is thought to function as a transcription factor, playing a role in hematopoiesis, oncogenesis and immune responses. Multiple isoforms of ZBTB20 exist due to alternative splicing events.Specifications
| ZBTB20 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB20 | |
| DPZF, HOF, ODA 8S, ZBTB20, Zinc finger protein 288, ZNF288 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26137, 56490 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K0L9, Q9HC78 | |
| Zbtb20 | |
| ZBTB20 Fusion Protein Ag21140 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction