missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548862
This item is not returnable.
View return policy
Description
Predicted to react with mouse based on sequence homology.
May be involved in transcriptional regulation.
Specifications
| ZBTB22 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O15209 | |
| ZBTB22 | |
| KLH conjugated synthetic peptide between 476-502 amino acids from the C-terminal region of human ZBTB22. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ZBTB22 | |
| 1110008J20Rik; AI265210; AI415166; Bing1; fru; fruitless; Protein BING1; ZBTB22; ZBTB22A; Zfp297; zinc finger and BTB domain containing 22; zinc finger and BTB domain-containing protein 22; Zinc finger and BTB domain-containing protein 22A; zinc finger protein 297; ZNF297; ZNF297A | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 9278 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction