missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZBTB24 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15711591 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15711591 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15711591 Supplier Invitrogen™ Supplier No. PA553651

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QLQPYQLSTS GEQEIQLLVT DSVHNINFMP GPSQGISIVT AESSQNMTAD QAANLTLLTQ QPEQLQNLIL SAQQEQTEHI QSLNMIESQM GPSQTEPVHV ITLSKETLEH LHAHQEQTEE LHLATSTSDP AQH Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.

May be involved in BMP2-induced transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB24
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB24
Gene Accession No. O43167
Gene Alias BC055367; Bif1; BMP-induced factor 1; bone morphogenetic protein-induced factor 1; brain specific protein 1; Brain-specific protein 1; Bsg1; ICF2; KIAA0441; PATZ2; POZ (BTB) and AT hook containing zinc finger 2; Zbtb24; zinc finger and BTB domain containing 24; zinc finger and BTB domain-containing protein 24; Zinc finger protein 450; ZNF450
Gene Symbols ZBTB24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB24. Recombinant protein control fragment (Product #RP-91653).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9841
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.