missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZBTB24 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17888632
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17888632 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17888632 Supplier Invitrogen™ Supplier No. PA5142869

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_055612.2; NP_001157785.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

May be involved in BMP2-induced transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB24
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ZBTB24
Gene Accession No. O43167
Gene Alias BC055367; Bif1; BMP-induced factor 1; bone morphogenetic protein-induced factor 1; brain specific protein 1; Brain-specific protein 1; Bsg1; ICF2; KIAA0441; PATZ2; POZ (BTB) and AT hook containing zinc finger 2; Zbtb24; zinc finger and BTB domain containing 24; zinc finger and BTB domain-containing protein 24; Zinc finger protein 450; ZNF450
Gene Symbols ZBTB24
Host Species Goat
Immunogen Peptide with sequence C-PVEKDENYDPKTED.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9841
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.