missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598246
This item is not returnable.
View return policy
Description
ZBTB25 Polyclonal Antibody for Western Blot, IHC (P), ELISA
May be involved in transcriptional regulation. Classified as a putative transcription factor; human homolog is a DNA binding protein with two distantly spaced zinc fingers.
Specifications
| ZBTB25 | |
| Polyclonal | |
| Unconjugated | |
| Zbtb25 | |
| 2810462M08Rik; 2900064P18Rik; AI842128; C14orf51; KUP; Zbtb25; Zfp50; Zfp-50; zinc finger and BTB domain containing 25; zinc finger and BTB domain-containing protein 25; Zinc finger protein 46; zinc finger protein 50; zinc finger protein KUP; ZNF46 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7597 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24278 | |
| Zbtb25 | |
| Recombinant Human Zinc finger and BTB domain-containing protein 25 protein (266-435AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction