missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB38 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18509AF488
This item is not returnable.
View return policy
Description
ZBTB38 Polyclonal antibody specifically detects ZBTB38 in Human samples. It is validated for Western Blot
Specifications
| ZBTB38 | |
| Polyclonal | |
| Western Blot | |
| CIBZ, FLJ22332, FLJ31131, FLJ35036, zinc finger and BTB domain containing 38, zinc finger and BTB domain-containing protein 38, ZNF921 | |
| Recognizes region between residues Between between 200 and 250.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 253461 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction