missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB4 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76517DL594
This item is not returnable.
View return policy
Description
ZBTB4 Polyclonal antibody specifically detects ZBTB4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ZBTB4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| KAISO-L1, zinc finger and BTB domain containing 4 | |
| Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human ZBTB4. The immunogen is located within amino acids 790 - 840 of ZBTB4. Amino Acid Squence: AGTRPGDVKEEAPQEMQV | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 57659 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction