missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: GEKEHMRVVV KSEPLSSPEP QDEVSDVTSQ AEGSESVEVE GVVVSAEKID LSPESSDRSF SDPQSSTDRV GDIHILEVTN NLEHKSTFSI SNFLNKSRGN NFTANQNN Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB5 was identified though sequence analysis as a POZ domain Kruppel-like zinc finger (POK) protein. Further experiments indicated that it binds DNA and can directly repress transcription of the cell cycle arrest gene p21. ZBTB5 can also interact with co-repressor histone deacetylase complexes such as BCoR, NCoR, and SMRT via its POZ domain, resulting in deacetylation of histones Ac-H3 and Ac-H4 at the proximal promoter. ZBTB5 stimulates both cell proliferation and cell cycle progression, suggesting that it may act as a potential proto-oncogene.
Specifications
Specifications
| Antigen | ZBTB5 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZBTB5 |
| Gene Accession No. | O15062 |
| Gene Alias | 5930421I10; 9430083K24Rik; AI646847; expressed in brain and neural tube; Kiaa0354; mKIAA0354; RP11-397D12.5; Transcription factor ZNF-POZ; ZBTB5; zinc finger and BTB domain containing 5; zinc finger and BTB domain-containing protein 5 |
| Gene Symbols | ZBTB5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?